Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAAATCTCTCACAATTAAAAAT[A/G]GGTGGAAGGGCCAAATATAAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610650 MIM: 601033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADRM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADRM1 - adhesion regulating molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281437.1 | 11231 | Intron | NP_001268366.1 | |||
NM_001281438.1 | 11231 | Intron | NP_001268367.1 | |||
NM_007002.3 | 11231 | Intron | NP_008933.2 | |||
NM_175573.2 | 11231 | Intron | NP_783163.1 | |||
XM_005260257.1 | 11231 | Intron | XP_005260314.1 | |||
XM_011528503.1 | 11231 | Intron | XP_011526805.1 | |||
XM_017027602.1 | 11231 | Intron | XP_016883091.1 | |||
XM_017027603.1 | 11231 | Intron | XP_016883092.1 | |||
XM_017027604.1 | 11231 | Intron | XP_016883093.1 | |||
XM_017027605.1 | 11231 | Intron | XP_016883094.1 | |||
XM_017027606.1 | 11231 | Intron | XP_016883095.1 | |||
XM_017027607.1 | 11231 | Intron | XP_016883096.1 | |||
XM_017027608.1 | 11231 | Intron | XP_016883097.1 | |||
XM_017027609.1 | 11231 | Intron | XP_016883098.1 | |||
XM_017027610.1 | 11231 | Intron | XP_016883099.1 |
LAMA5 - laminin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005560.4 | 11231 | UTR 3 | NP_005551.3 | |||
XM_006723796.3 | 11231 | UTR 3 | XP_006723859.1 | |||
XM_006723798.3 | 11231 | Intron | XP_006723861.1 | |||
XM_011528818.2 | 11231 | UTR 3 | XP_011527120.1 | |||
XM_011528819.2 | 11231 | UTR 3 | XP_011527121.1 |