Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTACATTGCACAATATTTGAATAA[C/G]GTATATTTATTCAATGACCTAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612360 | ||||||||||||||||||||
Literature Links: |
ESF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESF1 - ESF1 nucleolar pre-rRNA processing protein homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF5 - NADH:ubiquinone oxidoreductase complex assembly factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039375.2 | 370 | Missense Mutation | AAC,AAG | N,K 109 | NP_001034464.1 | |
NM_024120.4 | 370 | Missense Mutation | AAC,AAG | N,K 109 | NP_077025.2 | |
XM_006723620.3 | 370 | Missense Mutation | AAC,AAG | N,K 109 | XP_006723683.1 | |
XM_006723622.2 | 370 | UTR 5 | XP_006723685.1 | |||
XM_006723623.2 | 370 | UTR 5 | XP_006723686.1 | |||
XM_006723624.2 | 370 | UTR 5 | XP_006723687.1 | |||
XM_011529342.2 | 370 | Missense Mutation | AAC,AAG | N,K 109 | XP_011527644.1 | |
XM_011529344.2 | 370 | Intron | XP_011527646.1 | |||
XM_017028061.1 | 370 | UTR 5 | XP_016883550.1 |