Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAGATCTGCAGCCTGGACTCTC[A/G]TTTCTGAACCTGGGCAGTGGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCMTD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCMTD2 - protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104925.1 | 449 | Silent Mutation | TCA,TCG | S,S 83 | NP_001098395.1 | |
NM_018257.2 | 449 | Silent Mutation | TCA,TCG | S,S 83 | NP_060727.2 |