Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTGCTTACAAACACGCCTCCT[C/T]CTGCAAGGCCCGGAGTGGACTCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607114 MIM: 600751 | ||||||||||||||||||||
Literature Links: |
ADAM33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM33 - ADAM metallopeptidase domain 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC1 - sialic acid binding Ig like lectin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023068.3 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | NP_075556.1 | |
XM_006723610.3 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_006723673.1 | |
XM_011529324.2 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_011527626.1 | |
XM_011529325.2 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_011527627.1 | |
XM_011529326.2 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_011527628.1 | |
XM_011529327.2 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_011527629.1 | |
XM_011529328.2 | 5138 | Silent Mutation | AGA,AGG | R,R 1666 | XP_011527630.1 | |
XM_011529329.1 | 5138 | Intron | XP_011527631.1 | |||
XM_011529330.1 | 5138 | Silent Mutation | AGA,AGG | R,R 1580 | XP_011527632.1 | |
XM_011529331.1 | 5138 | Silent Mutation | AGA,AGG | R,R 1577 | XP_011527633.1 | |
XM_011529332.1 | 5138 | Silent Mutation | AGA,AGG | R,R 1463 | XP_011527634.1 | |
XM_011529333.2 | 5138 | Intron | XP_011527635.1 |