Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGTTGGATGAAGATAGTTTAA[C/T]CAAACAACCAGAAGAAGTATTTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604965 | ||||||||||||||||||||
Literature Links: |
STK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK4 - serine/threonine kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006282.3 | 131 | Missense Mutation | ACC,ATC | T,I 23 | NP_006273.1 | |
XM_005260530.2 | 131 | Missense Mutation | ACC,ATC | T,I 39 | XP_005260587.1 | |
XM_005260531.3 | 131 | Missense Mutation | ACC,ATC | T,I 39 | XP_005260588.1 | |
XM_005260532.3 | 131 | UTR 5 | XP_005260589.1 | |||
XM_005260533.2 | 131 | Missense Mutation | ACC,ATC | T,I 23 | XP_005260590.1 | |
XM_011529018.2 | 131 | UTR 5 | XP_011527320.1 | |||
XM_011529020.2 | 131 | Missense Mutation | ACC,ATC | T,I 39 | XP_011527322.1 | |
XM_017028029.1 | 131 | UTR 5 | XP_016883518.1 | |||
XM_017028030.1 | 131 | UTR 5 | XP_016883519.1 | |||
XM_017028031.1 | 131 | UTR 5 | XP_016883520.1 | |||
XM_017028032.1 | 131 | UTR 5 | XP_016883521.1 | |||
XM_017028033.1 | 131 | Missense Mutation | ACC,ATC | T,I 23 | XP_016883522.1 |
STK4-AS1 - STK4 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |