Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAGTTTGATTTGTGGCGAAA[C/T]GTTTTCTCACATTTGTTACATTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF334 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF334 - zinc finger protein 334 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270497.1 | 2443 | Silent Mutation | ACA,ACG | T,T 663 | NP_001257426.1 | |
NM_018102.4 | 2443 | Silent Mutation | ACA,ACG | T,T 664 | NP_060572.3 | |
XM_011528892.2 | 2443 | Silent Mutation | ACA,ACG | T,T 687 | XP_011527194.1 | |
XM_011528895.2 | 2443 | Silent Mutation | ACA,ACG | T,T 671 | XP_011527197.1 | |
XM_011528897.2 | 2443 | Silent Mutation | ACA,ACG | T,T 663 | XP_011527199.1 | |
XM_017027933.1 | 2443 | Silent Mutation | ACA,ACG | T,T 687 | XP_016883422.1 | |
XM_017027934.1 | 2443 | Silent Mutation | ACA,ACG | T,T 687 | XP_016883423.1 | |
XM_017027935.1 | 2443 | Silent Mutation | ACA,ACG | T,T 671 | XP_016883424.1 | |
XM_017027936.1 | 2443 | Silent Mutation | ACA,ACG | T,T 664 | XP_016883425.1 | |
XM_017027937.1 | 2443 | Silent Mutation | ACA,ACG | T,T 687 | XP_016883426.1 | |
XM_017027938.1 | 2443 | Silent Mutation | ACA,ACG | T,T 664 | XP_016883427.1 | |
XM_017027939.1 | 2443 | Silent Mutation | ACA,ACG | T,T 663 | XP_016883428.1 | |
XM_017027940.1 | 2443 | Silent Mutation | ACA,ACG | T,T 663 | XP_016883429.1 | |
XM_017027941.1 | 2443 | Silent Mutation | ACA,ACG | T,T 663 | XP_016883430.1 | |
XM_017027942.1 | 2443 | Silent Mutation | ACA,ACG | T,T 659 | XP_016883431.1 | |
XM_017027943.1 | 2443 | Silent Mutation | ACA,ACG | T,T 659 | XP_016883432.1 | |
XM_017027944.1 | 2443 | Silent Mutation | ACA,ACG | T,T 626 | XP_016883433.1 |