Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CTCCTTACTCCTTTGAAAGAACGTC[C/T]AGACCTACCTCCTGTACAATATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610512 | ||||||||||||||||||||
Literature Links: |
SEC23B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC23B - Sec23 homolog B, coat complex II component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172745.1 | 607 | Missense Mutation | CCA,CTA | P,L 49 | NP_001166216.1 | |
NM_001172746.1 | 607 | Missense Mutation | CCA,CTA | P,L 49 | NP_001166217.1 | |
NM_006363.4 | 607 | Missense Mutation | CCA,CTA | P,L 49 | NP_006354.2 | |
NM_032985.4 | 607 | Missense Mutation | CCA,CTA | P,L 49 | NP_116780.1 | |
NM_032986.3 | 607 | Missense Mutation | CCA,CTA | P,L 49 | NP_116781.1 | |
XM_017027593.1 | 607 | Missense Mutation | CCA,CTA | P,L 49 | XP_016883082.1 |