Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTCATACTAACCTCTGTTGTTC[A/G]CTTTTTTGATATGCACTGAACTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYCP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYCP2 - synaptonemal complex protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014258.3 | 4553 | Silent Mutation | AGC,AGT | S,S 1456 | NP_055073.2 | |
XM_011528487.2 | 4553 | Silent Mutation | AGC,AGT | S,S 1463 | XP_011526789.1 | |
XM_011528488.2 | 4553 | Silent Mutation | AGC,AGT | S,S 1463 | XP_011526790.1 | |
XM_011528489.2 | 4553 | Silent Mutation | AGC,AGT | S,S 1456 | XP_011526791.1 | |
XM_011528490.2 | 4553 | Silent Mutation | AGC,AGT | S,S 1455 | XP_011526792.1 | |
XM_011528493.2 | 4553 | Silent Mutation | AGC,AGT | S,S 1291 | XP_011526795.1 | |
XM_017027586.1 | 4553 | Silent Mutation | AGC,AGT | S,S 1463 | XP_016883075.1 | |
XM_017027587.1 | 4553 | Silent Mutation | AGC,AGT | S,S 1462 | XP_016883076.1 | |
XM_017027588.1 | 4553 | Silent Mutation | AGC,AGT | S,S 1411 | XP_016883077.1 | |
XM_017027589.1 | 4553 | Silent Mutation | AGC,AGT | S,S 1406 | XP_016883078.1 | |
XM_017027590.1 | 4553 | Silent Mutation | AGC,AGT | S,S 1291 | XP_016883079.1 | |
XM_017027591.1 | 4553 | Silent Mutation | AGC,AGT | S,S 980 | XP_016883080.1 | |
XM_017027592.1 | 4553 | Intron | XP_016883081.1 |