Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGTGGCCGAGGTCACTGAGGGGG[C/T]CCGATTGTCTTTGTAGGCAGCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605608 | ||||||||||||||||||||
Literature Links: |
CLDN14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN14 - claudin 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146077.1 | 1138 | Missense Mutation | NP_001139549.1 | |||
NM_001146078.2 | 1138 | Missense Mutation | NP_001139550.1 | |||
NM_001146079.1 | 1138 | Missense Mutation | NP_001139551.1 | |||
NM_012130.3 | 1138 | Missense Mutation | NP_036262.1 | |||
NM_144492.2 | 1138 | Missense Mutation | NP_652763.1 |