Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCAGGGGGTTGGCGCGCAGGTC[C/G]GGCCGCTGCTTCAGGATTCCGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606806 MIM: 612412 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FTCD - formimidoyltransferase cyclodeaminase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATC1L - spermatogenesis and centriole associated 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142854.1 | 606 | Silent Mutation | CCC,CCG | P,P 289 | NP_001136326.1 | |
NM_032261.4 | 606 | Silent Mutation | CCC,CCG | P,P 135 | NP_115637.3 | |
XM_005261188.4 | 606 | Silent Mutation | CCC,CCG | P,P 289 | XP_005261245.1 | |
XM_011529756.2 | 606 | Silent Mutation | CCC,CCG | P,P 175 | XP_011528058.1 | |
XM_017028480.1 | 606 | Silent Mutation | CCC,CCG | P,P 289 | XP_016883969.1 |