Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGCAAATATGGCAAAATAGTA[T/C]GTGAAGGGTACAAGGGCATTTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613083 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTN1 - listerin E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320766.1 | 7654 | UTR 3 | NP_001307695.1 | |||
NM_015565.2 | 7654 | UTR 3 | NP_056380.2 | |||
XM_006723987.3 | 7654 | Intron | XP_006724050.1 | |||
XM_017028316.1 | 7654 | UTR 3 | XP_016883805.1 | |||
XM_017028317.1 | 7654 | UTR 3 | XP_016883806.1 |