Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAATTGCCTATAATGCTTCTCTC[A/G]TTGAGTTGGAGTCCCAAAACCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC28 - DnaJ heat shock protein family (Hsp40) member C28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040192.2 | 897 | Nonsense Mutation | CGA,TGA | R,* 150 | NP_001035282.1 | |
NM_001320746.1 | 897 | Nonsense Mutation | CGA,TGA | R,* 150 | NP_001307675.1 | |
NM_017833.4 | 897 | Nonsense Mutation | CGA,TGA | R,* 150 | NP_060303.2 |
TMEM50B - transmembrane protein 50B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |