Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAAAATGTAGCATAAAGGTTTA[A/G]GTCCTGGGATACTGAATTTATAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616485 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB21 - zinc finger and BTB domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098402.1 | 5727 | UTR 3 | NP_001091872.1 | |||
NM_001098403.1 | 5727 | UTR 3 | NP_001091873.1 | |||
NM_001320729.1 | 5727 | UTR 3 | NP_001307658.1 | |||
NM_001320731.1 | 5727 | UTR 3 | NP_001307660.1 | |||
NM_020727.4 | 5727 | UTR 3 | NP_065778.3 | |||
XM_005261121.3 | 5727 | UTR 3 | XP_005261178.1 | |||
XM_011529588.2 | 5727 | UTR 3 | XP_011527890.1 | |||
XM_011529590.2 | 5727 | UTR 3 | XP_011527892.1 | |||
XM_017028360.1 | 5727 | UTR 3 | XP_016883849.1 | |||
XM_017028361.1 | 5727 | UTR 3 | XP_016883850.1 |