Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCAGCAGGAGGTCATCTCGCC[A/G]GACACTACCGAGAGAAACAACAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120240 | ||||||||||||||||||||
Literature Links: |
COL6A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL6A2 - collagen type VI alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001849.3 | 457 | Silent Mutation | CCA,CCG | P,P 28 | NP_001840.3 | |
NM_058174.2 | 457 | Silent Mutation | CCA,CCG | P,P 28 | NP_478054.2 | |
NM_058175.2 | 457 | Silent Mutation | CCA,CCG | P,P 28 | NP_478055.2 | |
XM_011529451.1 | 457 | Silent Mutation | CCA,CCG | P,P 28 | XP_011527753.1 |