Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGGTCAATAGCATTTCAGAGG[G/T]CTGCTACCACTGCACAAATACCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRTAP24-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRTAP24-1 - keratin associated protein 24-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085455.2 | 773 | Missense Mutation | AGA,AGC | R,S 249 | NP_001078924.1 |
KRTAP25-1 - keratin associated protein 25-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |