Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAATGGCCTCCTCTTGGTATTTC[C/T]TCTGCTGTTGATTTTTTGCATAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605938 MIM: 602259 | ||||||||||||||||||||
Literature Links: |
LOC105372795 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105372795 - uncharacterized LOC105372795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGP - phosphatidylinositol glycan anchor biosynthesis class P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320480.1 | 483 | Missense Mutation | AAG,AGG | K,R 101 | NP_001307409.1 | |
NM_016430.3 | 483 | Missense Mutation | AAG,AGG | K,R 75 | NP_057514.2 | |
NM_153681.2 | 483 | Missense Mutation | AAG,AGG | K,R 125 | NP_710148.1 | |
NM_153682.2 | 483 | Missense Mutation | AAG,AGG | K,R 101 | NP_710149.1 | |
XM_005260990.4 | 483 | Intron | XP_005261047.1 | |||
XM_011529596.2 | 483 | Missense Mutation | AAG,AGG | K,R 101 | XP_011527898.1 | |
XM_017028364.1 | 483 | Missense Mutation | AAG,AGG | K,R 101 | XP_016883853.1 | |
XM_017028365.1 | 483 | Missense Mutation | AAG,AGG | K,R 75 | XP_016883854.1 |
TTC3 - tetratricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |