Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCAGGAGTCACAGATGAGAGC[A/G]GAGATCCAGGACATGAAGCAGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602915 | ||||||||||||||||||||
Literature Links: |
WRB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WRB - tryptophan rich basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146218.1 | 294 | Silent Mutation | GCA,GCG | A,A 17 | NP_001139690.1 | |
NM_004627.4 | 294 | Silent Mutation | GCA,GCG | A,A 51 | NP_004618.2 |
WRB-SH3BGR - WRB-SH3BGR readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317744.1 | 294 | Silent Mutation | GCA,GCG | A,A 51 | NP_001304673.1 |