Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGTTTATATCTCTAAATGAAT[A/G]TGTTTTAAAATGTAAAAATGGTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RWDD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RWDD2B - RWD domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP16 - ubiquitin specific peptidase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001992.1 | Intron | NP_001001992.1 | ||||
NM_001032410.1 | Intron | NP_001027582.1 | ||||
NM_006447.2 | Intron | NP_006438.1 | ||||
XM_017028257.1 | Intron | XP_016883746.1 | ||||
XM_017028258.1 | Intron | XP_016883747.1 | ||||
XM_017028259.1 | Intron | XP_016883748.1 | ||||
XM_017028260.1 | Intron | XP_016883749.1 | ||||
XM_017028261.1 | Intron | XP_016883750.1 |