Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTATCTGAGTTTTTCATCTTTA[A/T]ATCTAAAAATCTAATCTGAAAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611428 MIM: 182465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DONSON PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DONSON - downstream neighbor of SON | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017613.3 | 1940 | UTR 3 | NP_060083.1 |
SON - SON DNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291411.1 | 1940 | Intron | NP_001278340.1 | |||
NM_001291412.1 | 1940 | Intron | NP_001278341.1 | |||
NM_032195.2 | 1940 | Intron | NP_115571.2 | |||
NM_138927.2 | 1940 | Intron | NP_620305.2 |