Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAGCAAACTTGGGGTTCATGAC[C/T]GTCGTGGTGGCGCTCTTGAAAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600065 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGB2 - integrin subunit beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000211.4 | 2530 | Silent Mutation | ACA,ACG | T,T 760 | NP_000202.3 | |
NM_001127491.2 | 2530 | Silent Mutation | ACA,ACG | T,T 760 | NP_001120963.2 | |
NM_001303238.1 | 2530 | Silent Mutation | ACA,ACG | T,T 691 | NP_001290167.1 | |
XM_006724001.1 | 2530 | Silent Mutation | ACA,ACG | T,T 691 | XP_006724064.1 | |
XM_017028341.1 | 2530 | Silent Mutation | ACA,ACG | T,T 691 | XP_016883830.1 |