Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGCCAGCCTGGTGTCCAGCCGCC[C/G]GTCGTCGCCAGAGCCCGATGACCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606386 | ||||||||||||||||||||
Literature Links: |
OLIG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLIG2 - oligodendrocyte lineage transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005806.3 | 423 | Missense Mutation | CCG,CGG | P,R 12 | NP_005797.1 | |
XM_005260908.1 | 423 | Missense Mutation | CCG,CGG | P,R 12 | XP_005260965.1 |