Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTAAGTACAATATTATACCTG[C/T]AAAGTGGTGGTCTGAGAAAGAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609252 | ||||||||||||||||||||
Literature Links: |
LIPI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIPI - lipase I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302998.1 | 1335 | Missense Mutation | TAC,TGC | Y,C 436 | NP_001289927.1 | |
NM_001302999.1 | 1335 | Missense Mutation | TAC,TGC | Y,C 406 | NP_001289928.1 | |
NM_001303000.1 | 1335 | Missense Mutation | TAC,TGC | Y,C 430 | NP_001289929.1 | |
NM_001303001.1 | 1335 | Missense Mutation | ACA,GCA | T,A 340 | NP_001289930.1 | |
NM_198996.3 | 1335 | Missense Mutation | TAC,TGC | Y,C 457 | NP_945347.2 | |
XM_006723965.3 | 1335 | Intron | XP_006724028.1 |