Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGAGAGGGAAGGAAACTGGCC[C/T]CGGGAAGGCGGCTTCAGGCCACGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603191 MIM: 171860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C21orf2 - chromosome 21 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271440.1 | 701 | Silent Mutation | CGA,CGG | R,R 195 | NP_001258369.1 | |
NM_001271441.1 | 701 | Silent Mutation | CGA,CGG | R,R 195 | NP_001258370.1 | |
NM_001271442.1 | 701 | Silent Mutation | CGA,CGG | R,R 154 | NP_001258371.1 | |
NM_004928.2 | 701 | Silent Mutation | CGA,CGG | R,R 196 | NP_004919.1 | |
XM_006724051.3 | 701 | Silent Mutation | CGA,CGG | R,R 221 | XP_006724114.1 | |
XM_006724053.3 | 701 | Silent Mutation | CGA,CGG | R,R 88 | XP_006724116.1 | |
XM_017028470.1 | 701 | Silent Mutation | CGA,CGG | R,R 263 | XP_016883959.1 | |
XM_017028471.1 | 701 | Silent Mutation | CGA,CGG | R,R 179 | XP_016883960.1 | |
XM_017028472.1 | 701 | Silent Mutation | CGA,CGG | R,R 87 | XP_016883961.1 |
PFKL - phosphofructokinase, liver type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |