Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTCCAACCCATTTTCCAACAGA[C/T]TATCTGCATGGTCCACTTTTATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00515 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00515 - long intergenic non-protein coding RNA 515 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL39 - mitochondrial ribosomal protein L39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017446.3 | Intron | NP_059142.2 | ||||
NM_080794.3 | Intron | NP_542984.2 | ||||
XM_006724026.3 | Intron | XP_006724089.1 | ||||
XM_011529651.2 | Intron | XP_011527953.1 |