Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTCTATTTCAACTCTGCTTTC[A/G]AGATTAAAAAGCTCTTTATCTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIS18A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIS18A - MIS18 kinetochore protein A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018944.2 | 636 | Silent Mutation | CTC,CTT | L,L 195 | NP_061817.1 | |
XM_017028400.1 | 636 | Intron | XP_016883889.1 | |||
XM_017028401.1 | 636 | Intron | XP_016883890.1 | |||
XM_017028402.1 | 636 | Intron | XP_016883891.1 |