Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTCTTTTCAAGATTGTAAGAA[A/T]GGCAGAATTTCATGAGCCACTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604735 | ||||||||||||||||||||
Literature Links: |
USP16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP16 - ubiquitin specific peptidase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001992.1 | Intron | NP_001001992.1 | ||||
NM_001032410.1 | Intron | NP_001027582.1 | ||||
NM_006447.2 | Intron | NP_006438.1 | ||||
XM_017028257.1 | Intron | XP_016883746.1 | ||||
XM_017028258.1 | Intron | XP_016883747.1 | ||||
XM_017028259.1 | Intron | XP_016883748.1 | ||||
XM_017028260.1 | Intron | XP_016883749.1 | ||||
XM_017028261.1 | Intron | XP_016883750.1 |