Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTCATTGATTGGCTGCCCGTC[A/G]AACCTGAATCTGATCTGCCTCATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602231 MIM: 603124 | ||||||||||||||||||||
Literature Links: |
LINC01424 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01424 - long intergenic non-protein coding RNA 1424 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUMO3 - small ubiquitin-like modifier 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286416.1 | 344 | Silent Mutation | NP_001273345.1 | |||
NM_006936.2 | 344 | Silent Mutation | NP_008867.2 |
UBE2G2 - ubiquitin conjugating enzyme E2 G2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |