Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAAGAAGCTGAAAGCCCACAAA[C/T]GTAAGTCTGAACTAGGGAAGCCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609196 | ||||||||||||||||||||
Literature Links: |
LOC105372779 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105372779 - uncharacterized LOC105372779 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRAP - melanocortin 2 receptor accessory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285394.1 | 293 | Intron | NP_001272323.1 | |||
NM_178817.3 | 293 | Missense Mutation | CAT,TAT | H,Y 36 | NP_848932.1 | |
NM_206898.1 | 293 | Missense Mutation | CAT,TAT | H,Y 36 | NP_996781.1 | |
XM_006724028.3 | 293 | Missense Mutation | CAT,TAT | H,Y 36 | XP_006724091.1 | |
XM_017028407.1 | 293 | Missense Mutation | CAT,TAT | H,Y 36 | XP_016883896.1 |