Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTAGAGGTCGAGAGTCCTCTC[A/G]TGGGGGTCTCCATGTTCAAGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607358 MIM: 171860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIRE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIRE - autoimmune regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFKL - phosphofructokinase, liver type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002021.2 | 282 | Missense Mutation | CAT,CGT | H,R 15 | NP_001002021.2 | |
NM_002626.5 | 282 | Intron | NP_002617.3 | |||
XM_005261135.3 | 282 | Intron | XP_005261192.1 | |||
XM_005261137.4 | 282 | UTR 5 | XP_005261194.1 | |||
XM_006724011.1 | 282 | Missense Mutation | CAT,CGT | H,R 15 | XP_006724074.1 | |
XM_006724012.1 | 282 | Missense Mutation | CAT,CGT | H,R 15 | XP_006724075.1 | |
XM_011529603.2 | 282 | Intron | XP_011527905.1 | |||
XM_017028368.1 | 282 | Missense Mutation | CAT,CGT | H,R 15 | XP_016883857.1 | |
XM_017028369.1 | 282 | Intron | XP_016883858.1 | |||
XM_017028370.1 | 282 | UTR 5 | XP_016883859.1 | |||
XM_017028371.1 | 282 | UTR 5 | XP_016883860.1 | |||
XM_017028372.1 | 282 | UTR 5 | XP_016883861.1 |