Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAGTCCTGGAGGTCTGACTGTC[C/T]AGTTTCTTCTTCTTCAGAATTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSPH1 - radial spoke head 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286506.1 | 1075 | Missense Mutation | AGA,GGA | R,G 265 | NP_001273435.1 | |
NM_080860.3 | 1075 | Missense Mutation | AGA,GGA | R,G 303 | NP_543136.1 | |
XM_005261208.2 | 1075 | Missense Mutation | AGA,GGA | R,G 234 | XP_005261265.1 | |
XM_011529786.1 | 1075 | Missense Mutation | AGA,GGA | R,G 279 | XP_011528088.1 |