Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCATGGCTGTGATGTAATTTTTC[A/G]TTAGTTGTAATTAACAATCTTCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPI - lipase I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM11 - RNA binding motif protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320602.1 | Intron | NP_001307531.1 | ||||
NM_144770.4 | Intron | NP_658983.3 | ||||
XM_005260997.4 | Intron | XP_005261054.1 | ||||
XM_017028384.1 | Intron | XP_016883873.1 | ||||
XM_017028385.1 | Intron | XP_016883874.1 | ||||
XM_017028386.1 | Intron | XP_016883875.1 | ||||
XM_017028387.1 | Intron | XP_016883876.1 |