Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAACTGGATGGCAGGTACATTT[C/T]ATTCTGATTCTTTCTTTATCGTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602490 | ||||||||||||||||||||
Literature Links: |
NRIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRIP1 - nuclear receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003489.3 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | NP_003480.2 | |
XM_005261063.3 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_005261120.1 | |
XM_005261065.3 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_005261122.1 | |
XM_011529747.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_011528049.1 | |
XM_011529748.2 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_011528050.1 | |
XM_011529749.2 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_011528051.1 | |
XM_011529751.2 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_011528053.1 | |
XM_011529752.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_011528054.1 | |
XM_017028473.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_016883962.1 | |
XM_017028474.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_016883963.1 | |
XM_017028475.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_016883964.1 | |
XM_017028476.1 | 4293 | Silent Mutation | TAA,TGA | *,* 1159 | XP_016883965.1 |