Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTCTCACAGTCCCAGAGACTT[C/T]TGGACGATCTGCTTGGCAATCTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603124 | ||||||||||||||||||||
Literature Links: |
UBE2G2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2G2 - ubiquitin conjugating enzyme E2 G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202489.1 | 486 | Silent Mutation | CAA,CAG | Q,Q 90 | NP_001189418.1 | |
NM_003343.5 | 486 | Silent Mutation | CAA,CAG | Q,Q 160 | NP_003334.2 | |
NM_182688.2 | 486 | Silent Mutation | CAA,CAG | Q,Q 132 | NP_872630.1 | |
XM_017028469.1 | 486 | Silent Mutation | CAA,CAG | Q,Q 132 | XP_016883958.1 |