Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCAGAATTTCTCAAAAGAAATA[C/T]TAAAAATCAGAAACATATACCATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGAP3 - ADP ribosylation factor GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142293.1 | 1912 | UTR 3 | NP_001135765.1 | |||
NM_014570.4 | 1912 | UTR 3 | NP_055385.3 | |||
XM_005261525.3 | 1912 | UTR 3 | XP_005261582.1 |