Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGGGCGCTCTGCACATCTGA[C/T]GGCCGCTTCTTCTGCACGCCCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604293 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DENND6B - DENN domain containing 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001794.3 | 1296 | Silent Mutation | CCA,CCG | P,P 408 | NP_001001794.3 | |
XM_005261914.3 | 1296 | Silent Mutation | CCA,CCG | P,P 457 | XP_005261971.1 | |
XM_011530692.2 | 1296 | Silent Mutation | CCA,CCG | P,P 457 | XP_011528994.1 | |
XM_011530693.2 | 1296 | Silent Mutation | CCA,CCG | P,P 408 | XP_011528995.1 | |
XM_017028801.1 | 1296 | Silent Mutation | CCA,CCG | P,P 201 | XP_016884290.1 |
PLXNB2 - plexin B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |