Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAGCTCCAGCCACAGCAGCTGC[C/T]GCTCCCGATGGGTGCCCTTCATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601279 MIM: 606810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGCR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGCR6 - DiGeorge syndrome critical region gene 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRODH - proline dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195226.1 | Intron | NP_001182155.1 | ||||
NM_016335.4 | Intron | NP_057419.4 |