Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACAGCATTGGCTCCACCACAG[C/T]GTGGTACAGAGCCTGGTAACCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf31 - chromosome 22 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015370.1 | 577 | Missense Mutation | ACT,GCT | T,A 219 | NP_056185.1 | |
XM_005261491.3 | 577 | Missense Mutation | ACT,GCT | T,A 115 | XP_005261548.1 | |
XM_011530096.2 | 577 | Missense Mutation | ACT,GCT | T,A 178 | XP_011528398.1 | |
XM_017028741.1 | 577 | Missense Mutation | ACT,GCT | T,A 178 | XP_016884230.1 |
ZNRF3 - zinc and ring finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |