Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGAGTGAGCGGGTCATCACGTC[C/T]CGTCAGGGGCCGCACATCCGCGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607422 MIM: 142708 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCAT - glycine C-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171690.1 | 212 | Silent Mutation | TCC,TCT | S,S 52 | NP_001165161.1 | |
NM_014291.3 | 212 | Intron | NP_055106.1 | |||
XM_005261409.3 | 212 | Intron | XP_005261466.1 | |||
XM_005261410.3 | 212 | Intron | XP_005261467.1 | |||
XM_005261411.1 | 212 | Intron | XP_005261468.1 | |||
XM_011530023.1 | 212 | Intron | XP_011528325.1 | |||
XM_017028674.1 | 212 | Intron | XP_016884163.1 | |||
XM_017028675.1 | 212 | Intron | XP_016884164.1 | |||
XM_017028676.1 | 212 | Intron | XP_016884165.1 | |||
XM_017028677.1 | 212 | Silent Mutation | TCC,TCT | S,S 52 | XP_016884166.1 |
H1F0 - H1 histone family member 0 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005318.3 | 212 | Intron | NP_005309.1 |