Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGACCTTGTGTACCCGCTTGA[C/T]GCTGTGGTGCTGCCAGGCGGTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100652901 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100652901 - uncharacterized LOC100652901 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRP5L - LDL receptor related protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135772.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | NP_001129244.1 | |
NM_182492.2 | 3682 | Intron | NP_872298.1 | |||
XM_006724372.3 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_006724435.1 | |
XM_017029083.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884572.1 | |
XM_017029084.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884573.1 | |
XM_017029085.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884574.1 | |
XM_017029086.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884575.1 | |
XM_017029087.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884576.1 | |
XM_017029088.1 | 3682 | Missense Mutation | ATC,GTC | I,V 186 | XP_016884577.1 |