Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAACAGTCACTCCTGGCATTCC[A/G]TGGCTGAGAACATTCAGGAAGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SELO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SELO - selenoprotein O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRABD - TraB domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320484.1 | Intron | NP_001307413.1 | ||||
NM_001320485.1 | Intron | NP_001307414.1 | ||||
NM_001320487.1 | Intron | NP_001307416.1 | ||||
NM_001320488.1 | Intron | NP_001307417.1 | ||||
NM_025204.3 | Intron | NP_079480.2 | ||||
XM_006724422.3 | Intron | XP_006724485.1 | ||||
XM_006724423.3 | Intron | XP_006724486.1 | ||||
XM_011530712.2 | Intron | XP_011529014.2 | ||||
XM_011530713.2 | Intron | XP_011529015.2 | ||||
XM_011530715.2 | Intron | XP_011529017.2 | ||||
XM_011530716.2 | Intron | XP_011529018.1 | ||||
XM_011530717.2 | Intron | XP_011529019.1 | ||||
XM_011530718.2 | Intron | XP_011529020.1 | ||||
XM_017028943.1 | Intron | XP_016884432.1 | ||||
XM_017028944.1 | Intron | XP_016884433.1 |