Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATCCTGGAGCTGGTGTCGCCTC[C/T]GCTGAAGCCTCCGCAAGTGCCGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606481 MIM: 611651 | ||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G3 - phospholipase A2 group III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015715.4 | 1629 | Missense Mutation | NP_056530.2 | |||
XM_011530204.1 | 1629 | Missense Mutation | XP_011528506.1 | |||
XM_011530205.1 | 1629 | Missense Mutation | XP_011528507.1 |