Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGCAGGCGCGCTTCTGCTGCC[A/G]CGTGGGCTGAGGGCACAGCACGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602049 MIM: 182453 | ||||||||||||||||||||
Literature Links: |
RAC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAC2 - ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002872.4 | 698 | Missense Mutation | CGG,TGG | R,W 183 | NP_002863.1 | |
XM_006724286.3 | 698 | Intron | XP_006724349.1 |
SSTR3 - somatostatin receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |