Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCCAAAGAAGGCATCCAAGCCC[A/G]AGAAGGCTGCCTCCAAAGCCCCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607422 MIM: 142708 | ||||||||||||||||||||
Literature Links: |
GCAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCAT - glycine C-acetyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
H1F0 - H1 histone family member 0 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005318.3 | 817 | Missense Mutation | AAG,GAG | K,E 127 | NP_005309.1 |