Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGAAGGCCTGGGCCAACACGGC[A/G]ATGATCTGCTTATACTTCCCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159540 | ||||||||||||||||||||
Literature Links: |
LIF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIF - leukemia inhibitory factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257135.1 | 741 | UTR 3 | NP_001244064.1 | |||
NM_002309.4 | 741 | Silent Mutation | ATC,ATT | I,I 195 | NP_002300.1 |
MGC20647 - uncharacterized protein MGC20647 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |