Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGGTCCCTTCTGCAAACCGCA[A/G]AACAAGTCATAAGCTTTCTTGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC5A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC5A4 - solute carrier family 5 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014227.2 | 1700 | Silent Mutation | TTC,TTT | F,F 606 | NP_055042.1 | |
XM_006724308.3 | 1700 | Silent Mutation | TTC,TTT | F,F 560 | XP_006724371.1 | |
XM_011530342.2 | 1700 | Silent Mutation | TTC,TTT | F,F 560 | XP_011528644.1 | |
XM_011530343.2 | 1700 | Silent Mutation | TTC,TTT | F,F 560 | XP_011528645.1 | |
XM_011530344.2 | 1700 | Silent Mutation | TTC,TTT | F,F 537 | XP_011528646.1 | |
XM_017028920.1 | 1700 | Silent Mutation | TTC,TTT | F,F 636 | XP_016884409.1 |