Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTGGGGGTGGAAGGTGGTCCC[A/G]GATCATCTCCTCATTCACGAAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613213 MIM: 611520 | ||||||||||||||||||||
Literature Links: |
CYB5R3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYB5R3 - cytochrome b5 reductase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000398.6 | 999 | Missense Mutation | CGG,TGG | R,W 259 | NP_000389.1 | |
NM_001129819.2 | 999 | Missense Mutation | CGG,TGG | R,W 236 | NP_001123291.1 | |
NM_001171660.1 | 999 | Missense Mutation | CGG,TGG | R,W 292 | NP_001165131.1 | |
NM_001171661.1 | 999 | Missense Mutation | CGG,TGG | R,W 236 | NP_001165132.1 | |
NM_007326.4 | 999 | Missense Mutation | CGG,TGG | R,W 236 | NP_015565.1 |
POLDIP3 - DNA polymerase delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU12 - RNA, U12 small nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |