Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTCTGATTACCTGTGCCTGAA[C/T]CCTGAACAGGGCCCTCACGTACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1I PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1I - calcium voltage-gated channel subunit alpha1 I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003406.1 | Intron | NP_001003406.1 | ||||
NM_021096.3 | Intron | NP_066919.2 | ||||
XM_017029035.1 | Intron | XP_016884524.1 | ||||
XM_017029036.1 | Intron | XP_016884525.1 | ||||
XM_017029037.1 | Intron | XP_016884526.1 |