Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATCCTTCCACAGTCTGCAAAC[C/T]AGTTCCTTCTGCAGTGTTTTCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOB2 - transducer of ERBB2, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016272.3 | 3304 | UTR 3 | NP_057356.1 | |||
XM_005261315.2 | 3304 | UTR 3 | XP_005261372.1 | |||
XM_006724105.3 | 3304 | UTR 3 | XP_006724168.1 | |||
XM_017028539.1 | 3304 | UTR 3 | XP_016884028.1 |