Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGGTCCTGATGGACCAGCAC[A/G]TGGGCTTTCTATGCAACGAGGTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607110 | ||||||||||||||||||||
Literature Links: |
APOBEC3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC3B - apolipoprotein B mRNA editing enzyme catalytic subunit 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270411.1 | 758 | Missense Mutation | ATG,GTG | M,V 235 | NP_001257340.1 | |
NM_004900.4 | 758 | Missense Mutation | ATG,GTG | M,V 235 | NP_004891.4 |
APOBEC3B-AS1 - APOBEC3B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |