Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGTGCTCAGATCTTCATGTAA[C/T]CTAGCGATCGCACGTTTTACCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ENTHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENTHD1 - ENTH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152512.3 | 1965 | Silent Mutation | NP_689725.2 | |||
XM_006724149.3 | 1965 | Silent Mutation | XP_006724212.1 | |||
XM_006724150.3 | 1965 | Silent Mutation | XP_006724213.1 | |||
XM_006724151.1 | 1965 | Missense Mutation | XP_006724214.1 | |||
XM_006724153.3 | 1965 | Intron | XP_006724216.1 | |||
XM_011529927.2 | 1965 | Silent Mutation | XP_011528229.1 | |||
XM_011529928.2 | 1965 | Silent Mutation | XP_011528230.1 | |||
XM_011529930.2 | 1965 | Silent Mutation | XP_011528232.1 | |||
XM_011529931.2 | 1965 | Intron | XP_011528233.1 |